7-Ethoxycumarin

Lieferant: Apollo Scientific

Synonyme: 7-Ethoxy-2H-chromen-2-one

BIE1002-1G BIE1002-5G
APOSBIE1002-1GEA 24.8 EUR
APOSBIE1002-1G APOSBIE1002-5G
7-Ethoxycumarin
7-Ethoxycumarin

Substrate for CYP2B6.

Formel: C₁₁H₁₀O₃
Molecular Weight: 190,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006877
CAS-Nummer: 31005-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR