N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid
Lieferant: Apollo Scientific
Gefahr
Synonyme: 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimid Hydrochlorid, EDAC HCl, WSC HCl, EDAC Hydrochlorid, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Monohydrochlorid, N'-(Ethylkohlenstoffimidoyl)-N,N-dimethylpropan-1,3-diaminmonohydrochlorid, EDC Hydrochlorid, EDC HCl
BIE1308-100G BIE1308-500G BIE1308-1KG
APOSBIE1308-100GEA 26.1 EUR
APOSBIE1308-100G APOSBIE1308-500G APOSBIE1308-1KG
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid
EDAC is a water soluble condensing reagent.
Formel: C₈H₁₇N₃·HCl
Molecular Weight: 191.7 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00012503
CAS-Nummer: 25952-53-8
EINECS: 247-361-2