all cis-5,8,11,14,17-Eicosapentaenoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5(Z),8(Z),11(Z),14(Z),17(Z)-Eicosapentaenoic Acid, Timnodonic acid, cis-5,8,11,14,17-Eicosapentaenoic acid, EPA

BIE1310-5G BIE1310-10G BIE1310-25G
APOSBIE1310-5GEA 109 EUR
APOSBIE1310-5G APOSBIE1310-10G APOSBIE1310-25G
all cis-5,8,11,14,17-Eicosapentaenoic acid
all cis-5,8,11,14,17-Eicosapentaenoic acid
Formel: C₂₀H₃₀O₂
Molecular Weight: 302,46 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00065716
CAS-Nummer: 10417-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR