Elacridar
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-[4-[2-(3,4-Dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)ethyl]phenyl]-9,10-dihydro-5-methoxy-9-oxo-4-acridinecarboxamide, N-(4-(2-(6,7-Dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)ethyl)phenyl)-5-methoxy-9-oxo-9,10-dihydroacridine-4-carboxamide
BIE3025-25MG
BIE3025-50MG
BIE3025-10MG
APOSBIE3025-25MGEA
139
EUR
APOSBIE3025-25MG
APOSBIE3025-50MG
APOSBIE3025-10MG
Elacridar
Elacridar
Formel:
C₃₄H₃₃N₃O₅ Molecular Weight: 563.654 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00912604 CAS-Nummer: 143664-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...