5-Fluoruracil

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,4-Dihydroxy-5-fluorpyrimidin, Fluoruracil, 5-FU

BIF0123-25G BIF0123-50G BIF0123-5G
APOSBIF0123-25GEA 69.6 EUR
APOSBIF0123-25G APOSBIF0123-50G APOSBIF0123-5G
5-Fluoruracil
5-Fluoruracil

This pyrimidine analogue, is an antineoplastic agent.

Formel: C₄H₃FN₂O₂
Molecular Weight: 130.08 g/mol
Siedepunkt: 190…200 °C/0.1mmHg
Schmelzpunkt: 282…286 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00006018
CAS-Nummer: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1 III
Merck Index: 13,04208

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR