Tacrolimus

Lieferant: Apollo Scientific
Achtung

BIF1003-250MG BIF1003-1G
APOSBIF1003-250MGEA 132 EUR
APOSBIF1003-250MG APOSBIF1003-1G
Tacrolimus
Tacrolimus
Formel: C₄₄H₆₉NO₁₂
Lagertemperatur: Gefrierschrank
MDL: MFCD00869853
CAS-Nummer: 104987-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR