Fumonisin B2 (aus Fusarium moniliforme)
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1,2,3-Propanetricarboxylic acid, 1,1?-[1-(12-amino-4,11-dihydroxy-2-methyl-tridecyl)-2-(1-methylpentyl)-1,2-ethane-diyl]ester, Fumonisin B2 from Fusarium moniliforme, sphingosine N-acyltransferase inhibitor, 1,2,3-Propanetricarboxylic acid, 1,1'-(1-(2-amino-9,11-dihydroxy-2-methyltridecyl)-2-(1-methylpentyl)-1,2-ethanediyl) ester
BIF1005-1MG
BIF1005-100UG
APOSBIF1005-1MGEA
551
EUR
APOSBIF1005-1MG
APOSBIF1005-100UG
Fumonisin B2 (aus Fusarium moniliforme)
Fumonisin B2
Formel:
C₃₄H₅₉NO₁₄ Molecular Weight: 705,839 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00133350 CAS-Nummer: 116355-84-1 UN: 2811 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...