Fmoc-D-Lys(Dde)-OH 97%

Lieferant: Apollo Scientific
Achtung

BIF1011-1G BIF1011-5G
APOSBIF1011-1GEA 145 EUR
APOSBIF1011-1G APOSBIF1011-5G
Fmoc-D-Lys(Dde)-OH 97%
Fmoc-D-Lys(Dde)-OH
Formel: C₃₁H₃₆N₂O₆
Molecular Weight: 532,6 g/mol
Schmelzpunkt: 76…78 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01862894
CAS-Nummer: 333973-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR