Forskolin (aus Coleus forskohlii)

Lieferant: Apollo Scientific
Achtung

Synonyme: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate

BIF1031-100MG BIF1031-25MG
APOSBIF1031-100MGEA 458 EUR
APOSBIF1031-100MG APOSBIF1031-25MG
Forskolin (aus Coleus forskohlii)
Forskolin

An activator of adenylate cyclase.

Formel: C₂₂H₃₄O₇
Molecular Weight: 410,51 g/mol
Schmelzpunkt: 228…230 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00082317
CAS-Nummer: 66575-29-9
EINECS: 266-410-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR