Fasudil hydrochloride
Lieferant: Apollo Scientific
Achtung
Synonyme: 5-(1-Homopiperazinyl)sulphonylisoquinoline Hydrochloride, 1-(5-Isoquinolinesulphonyl)homopiperazine Hydrochloride, Fasudil HCl, 5-((1,4-Diazepan-1-yl)sulphonyl)isoquinoline hydrochloride
BIF105-1G BIF105-250MG
APOSBIF105-1GEA 154 EUR
APOSBIF105-1G APOSBIF105-250MG
Fasudil hydrochloride
Fasudil hydrochloride
Kinase Inhibitor.
Formel: C₁₄H₁₇N₃O₂S
Molecular Weight: 327.83 g/mol
Schmelzpunkt: 222 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00943198
CAS-Nummer: 105628-07-7