Fluoresceindiacetat

Lieferant: Apollo Scientific
Achtung

Synonyme: Di-O-acetylfluorescein, 3,6-Diacetoxyfluoran, 3-Oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl diacetate

BIF4105-25G BIF4105-5G
APOSBIF4105-25GEA 132 EUR
APOSBIF4105-25G APOSBIF4105-5G
Fluoresceindiacetat
Fluoresceindiacetat
Formel: C₂₄H₁₆O₇
Molecular Weight: 416,38 g/mol
Schmelzpunkt: 200…203 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00005062
CAS-Nummer: 596-09-8
EINECS: 209-877-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR