PP242 freie Base ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)-1H-indol-5-ol, Torkinib, 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-1H-indol-5-ol
BIFK0004-10MG
BIFK0004-5MG
APOSBIFK0004-10MGEA
65.4
EUR
APOSBIFK0004-10MG
APOSBIFK0004-5MG
PP242 freie Base ≥99%
PP242
Formel:
C₁₆H₁₆N₆O Molecular Weight: 308,3 g/mol Schmelzpunkt: 146…156 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD12196869 CAS-Nummer: 1092351-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...