YM155 ≥98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Sepantronium Bromide, 1-(2-Methoxyethyl)-2-methyl-4,9-dioxo-3-(pyrazin-2-ylmethyl)-4,9-dihydro-1H-naphtho[2,3-d]imidazol-3-ium bromide

BIFK0009-50MG
APOSBIFK0009-50MGEA 139 EUR
APOSBIFK0009-50MG
YM155 ≥98%
YM155
Formel: C₂₀H₁₉BrN₄O₃
Molecular Weight: 443,29 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11983133
CAS-Nummer: 781661-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR