Afatinib freie Base ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
BIBW2992, Tovok
BIFK0010-25MG
BIFK0010-100MG
BIFK0010-250MG
APOSBIFK0010-25MGEA
87
EUR
APOSBIFK0010-25MG
APOSBIFK0010-100MG
APOSBIFK0010-250MG
Afatinib freie Base ≥99%
Afatinib
Formel:
C₂₄H₂₅ClFN₅O₃ Molecular Weight: 485,94 g/mol Schmelzpunkt: 130…142 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD12407405 CAS-Nummer: 850140-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...