Motesanib freie Base 99%
Lieferant: Apollo Scientific
Synonyme:
N-(3,3-Dimethylindolin-6-yl)-2-((pyridin-4-ylmethyl)amino)nicotinamide, AMG706
BIFK0029-10MG
BIFK0029-50MG
BIFK0029-100MG
APOSBIFK0029-10MGEA
65.4
EUR
APOSBIFK0029-10MG
APOSBIFK0029-50MG
APOSBIFK0029-100MG
Motesanib freie Base 99%
Motesanib
Formel:
C₂₂H₂₃N₅O Molecular Weight: 373.5 g/mol Schmelzpunkt: 140…151 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD10567689 CAS-Nummer: 453562-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...