PD-0173074 freie Base ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
PD 173074, 1-(tert-Butyl)-3-[7-[[4-(diethylamino)butyl]amino]-3-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-2-yl]urea, PD173074, 1-(tert-Butyl)-3-(2-((4-(diethylamino)butyl)amino)-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl)urea
BIFK0034-50MG
APOSBIFK0034-50MGEA
213
EUR
APOSBIFK0034-50MG
PD-0173074 freie Base ≥99%
PD-0173074
Formel:
C₂₈H₄₁N₇O₃ Molecular Weight: 523,7 g/mol Schmelzpunkt: 79…91 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD08705327 CAS-Nummer: 219580-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...