1,1-Dimethylpiperidin-1-ium-4-yloctadecyl phosphate ≥99%
Lieferant: Apollo Scientific
Synonyme:
1,1-Dimethylpiperidin-1-ium-4-yl octadecyl phosphate, D-21266, NSC 639966, KRX 41, Perifosine
BIFK0035-10MG
BIFK0035-1MG
BIFK0035-50MG
APOSBIFK0035-10MGEA
109
EUR
APOSBIFK0035-10MG
APOSBIFK0035-1MG
APOSBIFK0035-50MG
1,1-Dimethylpiperidin-1-ium-4-yloctadecyl phosphate ≥99%
1,1-Dimethylpiperidin-1-ium-4-yloctadecyl phosphate
Formel:
C₂₅H₅₂NO₄P Molecular Weight: 461,66 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00927554 CAS-Nummer: 157716-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...