StemRegenin 1 freie Base ≥99%

Lieferant: Apollo Scientific

Synonyme: 4-(2-((2-Benzo[b]thiphen-3-yl)-9-isopropyl-9H-purin-6-yl)amino)ethyl)phenol hydrochloride, SR1, 4-(2-((2-(Benzo[b]thiophen-3-yl)-9-isopropyl-9H-purin-6-yl)amino)ethyl)phenol

BIFK0042-10MG BIFK0042-50MG
APOSBIFK0042-10MGEA 72.5 EUR
APOSBIFK0042-10MG APOSBIFK0042-50MG
StemRegenin 1 freie Base ≥99%
StemRegenin 1
Formel: C₂₄H₂₃N₅OS
Molecular Weight: 429,542 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD16987861
CAS-Nummer: 1227633-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR