Vatalanib dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(4-Chlorophenyl)-4-(pyridin-4-ylmethyl)phthalazin-1-amine dihydrochloride, Vatalanib 2HCl

BIFK0044-25MG BIFK0044-10MG
APOSBIFK0044-25MGEA 82.8 EUR
APOSBIFK0044-25MG APOSBIFK0044-10MG
Vatalanib dihydrochloride
Vatalanib dihydrochloride
Formel: C₂₀H₁₇Cl₃N₄
Molecular Weight: 419,7348 g/mol
Schmelzpunkt: 284…286 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD08458964
CAS-Nummer: 212141-51-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR