AS-605240 freie Base ≥99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Quinoxalin-6-ylmethylene)thiazolidine-2,4-dione

BIFK0058-50MG
APOSBIFK0058-50MGEA 273 EUR
APOSBIFK0058-50MG
AS-605240 freie Base ≥99%
AS-605240
Formel: C₁₂H₇N₃O₂S
Molecular Weight: 257,2679 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11114066
CAS-Nummer: 648450-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR