L(-)-Xylose

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S,3R,4S)-2,3,4,5-Tetrahydroxypentanal, L-(-)-Xylose

BIFS0118-10G BIFS0118-5G
APOSBIFS0118-10GEA 79.9 EUR
APOSBIFS0118-10G APOSBIFS0118-5G
L(-)-Xylose
L(-)-Xylose
Formel: C₅H₁₀O₅
Molecular Weight: 150,13 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151096
CAS-Nummer: 609-06-3
EINECS: 210-174-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR