Glycitein
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one, 7,4'-Dihydroxy-6-methoxyisoflavone
BIG0110-10MG
BIG0110-5MG
APOSBIG0110-10MGEA
125
EUR
APOSBIG0110-10MG
APOSBIG0110-5MG
Glycitein
Glycitein
Isoflavone derived from Soybean.
Formel:
C₁₆H₁₂O₅ Molecular Weight: 284,27 g/mol Siedepunkt: 547,4 °C Schmelzpunkt: 330…340 °C Flammpunkt: 210 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00016679 CAS-Nummer: 40957-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...