N-(Phosphonomethyl)glycine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Glyphosat

BIG0158-1G BIG0158-5G
APOSBIG0158-1GEA 29 EUR
APOSBIG0158-1G APOSBIG0158-5G
N-(Phosphonomethyl)glycine 95%
N-(Phosphonomethyl)glycine

Inhibits the enzyme EPSPS which is involved in the shikimate pathway.

Formel: C₃H₈NO₅P
Molecular Weight: 169,07 g/mol
Schmelzpunkt: 230 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00055350
CAS-Nummer: 1071-83-6
EINECS: 213-997-4
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR