Fumagillin (aus Aspergillus fumigatus)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2E,4E,6E,8E)-10-(((3R,4S,5S,6R)-5-methoxy-4-((2R,3R)-2-methyl-3-(3-methylbut-2-en-1-yl)oxiran-2-yl)-1-oxaspiro[2.5]octan-6-yl)oxy)-10-oxodeca-2,4,6,8-tetraenoic acid
BIG0214-10MG
BIG0214-25MG
BIG0214-100MG
APOSBIG0214-10MGEA
79.9
EUR
APOSBIG0214-10MG
APOSBIG0214-25MG
APOSBIG0214-100MG
Fumagillin (aus Aspergillus fumigatus)
Fumagillin
Formel:
C₂₆H₃₄O₇ Molecular Weight: 458,551 g/mol Schmelzpunkt: 197…198 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD03990453 CAS-Nummer: 23110-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...