Penta-O-acetyl-β-D-galactopyranose 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: β-D-Galactose Pentaacetate, (2S,3R,4S,5S,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

BIG0303-100G BIG0303-1KG BIG0303-500G
APOSBIG0303-100GEA 52.2 EUR
APOSBIG0303-100G APOSBIG0303-1KG APOSBIG0303-500G
Penta-O-acetyl-β-D-galactopyranose 98%
Penta-O-acetyl-beta-D-galactopyranose
Formel: C₁₆H₂₂O₁₁
Molecular Weight: 390,34 g/mol
Schmelzpunkt: 139…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063259
CAS-Nummer: 4163-60-4
EINECS: 224-008-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR