Gibberellinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylen-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carbonsäure

BIG0500-10G BIG0500-5G
APOSBIG0500-10GEA 139 EUR
APOSBIG0500-10G APOSBIG0500-5G
Gibberellinsäure
Gibberellic acid

Gibberellic acid is a plant growth regulator.


  • Plant tissue culture tested

Formel: C₁₉H₂₂O₆
Molecular Weight: 346,38 g/mol
Schmelzpunkt: 227 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00079329
CAS-Nummer: 77-06-5
EINECS: 201-001-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR