Glycin (H-Gly-OH), ultrarein

Lieferant: Apollo Scientific
Achtung

Synonyme: Aminoacetic acid, Glycocoll, H-Gly-OH

BIG0503-1KG BIG0503-500G BIG0503-100G
APOSBIG0503-1KGEA 161 EUR
APOSBIG0503-1KG APOSBIG0503-500G APOSBIG0503-100G
Glycin (H-Gly-OH), ultrarein
Glycine

  • Plant tissue culture tested

Formel: NH₂CH₂COOH
Molecular Weight: 75,07 g/mol
Schmelzpunkt: 240…245 °C (Zersetzung)
Dichte: 1,595 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008131
CAS-Nummer: 56-40-6
EINECS: 200-272-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR