Gliotoxin (aus Gliocladium fimbriatum)
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,3,5a,6-Tetrahydro-6-hydroxy-3(hyroxymethyl)-2-methyl-10H-3a,10a-epidithio-pyrazinol[1,2α]indole-1,4-dione
BIG1003-1MG
BIG1003-5MG
APOSBIG1003-1MGEA
139
EUR
APOSBIG1003-1MG
APOSBIG1003-5MG
Gliotoxin (aus Gliocladium fimbriatum)
Gliotoxin
Formel:
C₁₃H₁₄N₂O₄S₂ Molecular Weight: 326,4 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
67-99-2 UN: 3462 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...