Guanosin ≥98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
BIG2502-100G
BIG2502-500G
APOSBIG2502-100GEA
79.9
EUR
APOSBIG2502-100G
APOSBIG2502-500G
Guanosin ≥98%
Guanosin
Formel:
C₁₀H₁₃N₅O₅ Molecular Weight: 283,24 g/mol Schmelzpunkt: 250 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010182 CAS-Nummer: 118-00-3 EINECS: 204-227-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...