Guanosin-5'-triphosphat Dinatriumsalz

Lieferant: Apollo Scientific
Achtung

Synonyme: ((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Guanosine-5'-triphosphate disodium salt, GTP disodium salt, Guanosine-5-triphosphoric aicd disodium salt

BIG4005-1G BIG4005-250MG BIG4005-500MG
APOSBIG4005-1GEA 109 EUR
APOSBIG4005-1G APOSBIG4005-250MG APOSBIG4005-500MG
Guanosin-5'-triphosphat Dinatriumsalz
Guanosin-5'-triphosphat Dinatriumsalz
Formel: C₁₀H₁₄N₅Na₂O₁₄P₃
Molecular Weight: 567.147 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03410297
CAS-Nummer: 56001-37-7
EINECS: 259-940-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR