D-Glucose-6-phosphate disodium salt

Lieferant: Apollo Scientific
Achtung

Synonyme: D-Glucose 6-phosphate disodium salt, Sodium (2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl phosphate, Robison Ester Disodium Salt

BIG4301-1G BIG4301-5G
APOSBIG4301-1GEA 58 EUR
APOSBIG4301-1G APOSBIG4301-5G
D-Glucose-6-phosphate disodium salt
D-Glucose-6-phosphate disodium salt
Formel: C₆H₁₁Na₂O₉P
Molecular Weight: 304.098 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00136037
CAS-Nummer: 3671-99-6
EINECS: 222-938-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR