Guanin ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-1H-purin-6(9H)-on, 2-Amino-6-oxopurin
BIG5137-100G
BIG5137-25G
APOSBIG5137-100GEA
50.9
EUR
APOSBIG5137-100G
APOSBIG5137-25G
Guanin ≥98%
Guanin
Formel:
C₅H₅N₅O Molecular Weight: 151,13 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071533 CAS-Nummer: 73-40-5 EINECS: 200-799-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...