Guanin Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-1H-purin-6(7H)-one hydrochloride
BIG5143-100G
BIG5143-25G
APOSBIG5143-100GEA
139
EUR
APOSBIG5143-100G
APOSBIG5143-25G
Guanin Hydrochlorid
Guanin Hydrochlorid
Formel:
C₅H₆ClN₅O Molecular Weight: 187,59 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00213670 CAS-Nummer: 635-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...