3-Hydroxy-2,4,6-tribrombenzoesäure ≥97%

Lieferant: Apollo Scientific

Synonyme: 2,4,6-Tribromo-3-hydroxybenzoic acid

BIH6066-1G BIH6066-25G BIH6066-5G
APOSBIH6066-1GEA 27.7 EUR
APOSBIH6066-1G APOSBIH6066-25G APOSBIH6066-5G
3-Hydroxy-2,4,6-tribrombenzoesäure ≥97%
3-Hydroxy-2,4,6-tribromobenzoic acid
Formel: C₇H₃Br₃O₃
Molecular Weight: 374.81 g/mol
Siedepunkt: 396
Schmelzpunkt: 146
Lagertemperatur: Raumtemperatur
MDL: MFCD00055557
CAS-Nummer: 14348-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR