N-Hydroxysuccinimid ≥97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Hydroxyazacyclopentan-2,5-dion, 1-Hydroxy-1,4-butandisäureimid, NHS, 1-Hydroxybernsteinsäureimid, 1-Hydroxy-2,5-pyrrolidindion, HOSu, N-Hydroxybernsteinsäureimid, N-Hydroxy-1,4-butandisäureimid
BIH6094-1KG
BIH6094-250G
BIH6094-100G
APOSBIH6094-1KGEA
122
EUR
APOSBIH6094-1KG
APOSBIH6094-250G
APOSBIH6094-100G
N-Hydroxysuccinimid ≥97%
N-Hydroxysuccinimid
Formel:
C₄H₅NO₃ Molecular Weight: 115.09 g/mol Siedepunkt: 363 Schmelzpunkt: 94…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005516 CAS-Nummer: 6066-82-6 EINECS: 228-001-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...