2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyltetrazoliumchlorid
Lieferant: Apollo Scientific
Achtung
Synonyme: 3-(4-Iodphenyl)-2-(4-nitrophenyl)-5-phenyltetrazoliumchlorid, 2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazoliumchlorid, INT, Iodnitrotetrazoliumchlorid, 3-(4-Iodphenyl)-2-(4-nitrophenyl)-5-phenyl-2H-tetrazoliumchlorid
BII7020-1G BII7020-500MG
APOSBII7020-1GEA 109 EUR
APOSBII7020-1G APOSBII7020-500MG
2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyltetrazoliumchlorid
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Used in colorimetric analysis of dehydrogenases
Formel: C₁₉H₁₃ClIN₅O₂
Molecular Weight: 505.7 g/mol
Schmelzpunkt: 245 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00011961
CAS-Nummer: 146-68-9