Jasmonic acid (mixture of isomers)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Carboxymethyl)-2-(2-pentenyl)cyclopentanone, 3-Oxo-2-(2-pentenyl)cyclopentaneacetic Acid
BIJ0379-100MG
BIJ0379-250MG
APOSBIJ0379-100MGEA
40.6
EUR
APOSBIJ0379-100MG
APOSBIJ0379-250MG
Jasmonic acid (mixture of isomers)
Jasmonic acid (mixture of isomers)
- Plant tissue culture tested
Formel:
C₁₂H₁₈O₃ Molecular Weight: 210,27 g/mol Dichte: 1,07 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
CAS-Nummer:
221682-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...