Apigenin 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-on, 4',5,7-Trihydroxyflavon, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyron, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-on

BIK9018-1G BIK9018-5G BIK9018-25G
APOSBIK9018-1GEA 43.5 EUR
APOSBIK9018-1G APOSBIK9018-5G APOSBIK9018-25G
Apigenin 97+%
Apigenin
Formel: C₁₅H₁₀O₅
Molecular Weight: 270.24 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006831
CAS-Nummer: 520-36-5
EINECS: 208-292-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR