Leptomycin A

Lieferant: Apollo Scientific

BIL2100-1MG BIL2100-5MG
APOSBIL2100-1MGEA 284 EUR
APOSBIL2100-1MG APOSBIL2100-5MG
Leptomycin A
Leptomycin A

  • Antibodies

Formel: C₃₂H₄₆O₆
Molecular Weight: 526,7 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 87081-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR