Leptomycin B

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-Ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-((2S,3S)-3-methyl-6-oxo-3,6-dihydro-2H-pyran-2-yl)-8-oxononadeca-2,10,12,16,18-pentaenoic acid

BIL2101-5MG BIL2101-10MG
APOSBIL2101-5MGEA 278 EUR
APOSBIL2101-5MG APOSBIL2101-10MG
Leptomycin B
Leptomycin B

Potent inhibitor of nuclear export.


Supplied as a 1mM solution in abs. ethanol

Formel: C₃₃H₄₈O₆
Molecular Weight: 540,73 g/mol
Flammpunkt: 11,1 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD06795848
CAS-Nummer: 87081-35-4
UN: 1170
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR