6-Mercaptopurine monohydrate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Purinethiol monohydrate, 6-Mercaptopurine hydrate, 7H-Purine-6-thiol hydrate, 6-Thiohypoxanthine monohydrate
BIM0129-25G
BIM0129-5G
APOSBIM0129-25GEA
46.4
EUR
APOSBIM0129-25G
APOSBIM0129-5G
6-Mercaptopurine monohydrate
6-Mercaptopurine monohydrate
6-Mercaptopurine is an antineoplastic agent which acts asan antimetabolite. It is an analogue of the natural purines hypoxanthine and adenine.
Formel:
C₅H₄N₄S·H₂O Molecular Weight: 170,19 g/mol (152,17 anhy) Schmelzpunkt: 315 °C (Zersetzung) Lagertemperatur: Kühlschrank |
MDL:
MFCD03854445 CAS-Nummer: 6112-76-1 EINECS: 200-037-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...