Salinomycin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(R)-2-((2R,5S,6R)-6-((2S,3S,4S,6R)-6-((2S,5S,7R,9S,10S,12R,15R)-2-((2R,5R,6S)-5-Ethyl-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)-15-hydroxy-2,10,12-trimethyl-1,6,8-trioxadispiro[4.1.57.35]pentadec-13-en-9-yl)-3-hydroxy-4-methyl-5-oxooctan-2-yl)-5-methyltetrahydro-2H-pyran-2-yl)butanoic acid
BIM0200-5MG
BIM0200-10MG
APOSBIM0200-5MGEA
122
EUR
APOSBIM0200-5MG
APOSBIM0200-10MG
Salinomycin
Salinomycin
Formel:
C₄₂H₇₀O₁₁ Molecular Weight: 751,00 g/mol Schmelzpunkt: 112,5…113.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD25541652 CAS-Nummer: 53003-10-4 UN: 3462 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...