Nifuratel
Lieferant: Apollo Scientific
Synonyme:
5-[(Methylthio)methyl]-3-[[(5-nitro-2-furanyl)methylene]amino]-2-oxazolidinone, 5-((Methylthio)methyl)-3-(((5-nitrofuran-2-yl)methylene)amino)oxazolidin-2-one
BIM0611-5G
BIM0611-25G
APOSBIM0611-5GEA
145
EUR
APOSBIM0611-5G
APOSBIM0611-25G
Nifuratel
Nifuratel
Formel:
C₁₀H₁₁N₃O₅S Molecular Weight: 285.27 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00057257 CAS-Nummer: 4936-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...