Maltoheptaose
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal
Activator of phosphorylase B for the preparation of heptulose-2-phosphate.
Formel:
C₄₂H₇₂O₃₆ Molecular Weight: 1152,9995 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00064605 CAS-Nummer: 34620-78-5 EINECS: 252-119-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...