(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide
Lieferant: Apollo Scientific
Achtung
Synonyme: 1-Fluoro-2,4-dinitrophenyl-5-L-alanine amide, N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, Propanamide, 2-[(5-fluoro-2,4-dinitrophenyl)amino]-, (2S)-
BIM106-100MG BIM106-250MG
APOSBIM106-100MGEA 21.9 EUR
APOSBIM106-100MG APOSBIM106-250MG
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide
Derivatised D- and L- amino acids can be resolved and quantified by HPLC. For reference see:Marfey, P. (1984) Carlsberg Res. Commun. 49, 591
Formel: C₉H₉FN₄O₅
Molecular Weight: 272,192 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00042049
CAS-Nummer: 95713-52-3