N-Acetylneuraminic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Neu5Ac, (4S,5R,6R,7S,8R)-5-Acetamido-4,6,7,8,9-pentahydroxy-2-oxononanoic acid, NANA, Sialic acid, 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-nonulopyranosonic acid, Lactaminic acid

BIM1062-25G BIM1062-5G
APOSBIM1062-25GEA 174 EUR
APOSBIM1062-25G APOSBIM1062-5G
N-Acetylneuraminic acid 98%
N-Acetylneuraminic acid
Formel: C₁₁H₁₉NO₉
Molecular Weight: 309,27 g/mol
Schmelzpunkt: 184…186 °C (Zers.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00006620
CAS-Nummer: 131-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR