Pyridoxal-5'-phosphat Monohydrat 98%

Lieferant: Apollo Scientific

Synonyme: Pyridoxal-5-phosphate hydrate, Pyridoxal 5-phosphate monohydrate, Pyridoxal 5'-phosphate monohydrate, (4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate hydrate

BIM1063-5G BIM1063-25G BIM1063-100G
APOSBIM1063-5GEA 36.4 EUR
APOSBIM1063-5G APOSBIM1063-25G APOSBIM1063-100G
Pyridoxal-5'-phosphat Monohydrat 98%
Pyridoxal-5'-phosphat Monohydrat
Formel: C₈H₁₀NO₆P·1H₂O
Molecular Weight: 265,17 g/mol (247,15 anhy)
Lagertemperatur: Gefrierschrank
MDL: MFCD00149414
CAS-Nummer: 41468-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR