4-Methylumbelliferyl-N-acetyl-β-D-galactosaminide hydrate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Methylumbelliferyl N-Acetyl-beta-D-galactosaminide, 4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-galactopyranoside
BIM1088-100MG
BIM1088-1G
BIM1088-250MG
APOSBIM1088-100MGEA
31.9
EUR
APOSBIM1088-100MG
APOSBIM1088-1G
APOSBIM1088-250MG
4-Methylumbelliferyl-N-acetyl-β-D-galactosaminide hydrate
4-Methylumbelliferyl-N-acetyl-beta-D-galactosaminide hydrate
Fluorescent substrate for N-acetyl-b-D-galactosaminidase.
Formel:
C₁₈H₂₁NO₈ Molecular Weight: 379,365 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00067658 CAS-Nummer: 36476-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...