4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside

Lieferant: Apollo Scientific

Synonyme: 4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside, 4-Methylumbelliferyl N-Acetyl-beta-D-glucosaminide

BIM1089-1G BIM1089-100MG BIM1089-500MG
APOSBIM1089-1GEA 261 EUR
APOSBIM1089-1G APOSBIM1089-100MG APOSBIM1089-500MG
4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside
4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside

Fluorescent substrate for N-acetyl-b-D-galactosaminidase.

Formel: C₁₈H₂₁NO₈
Molecular Weight: 379,365 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00065481
CAS-Nummer: 37067-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR