4-Methylumbelliferyl-alpha-D-galactopyranoside

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methylumbelliferyl-α-D-galactopyranoside, 4-methylumbelliferyl galactose, 4-MU-alpha-D-Gal, 4-methylumbelliferone-alpha-D-galactoside, 4-Methyl-7-(((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one, 4-methylumbelliferyl-galactopyranoside, ((beta-D)-galactosyloxy)-isomer

BIM1094-100MG BIM1094-250MG
APOSBIM1094-100MGEA 154 EUR
APOSBIM1094-100MG APOSBIM1094-250MG
4-Methylumbelliferyl-alpha-D-galactopyranoside
4-Methylumbelliferyl-alpha-D-galactopyranoside
Formel: C₁₆H₁₈O₈
Molecular Weight: 338,312 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00063278
CAS-Nummer: 38597-12-5
EINECS: 254-031-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR